Dextransulfat Natriumsalz, Calbiochem®

Lieferant: Merck Millipore Calbiochem

265152-10 265152-50
265152-10EA 118 EUR
265152-10 265152-50
Dextransulfat Natriumsalz, Calbiochem®
Dextran sulphate sodium salt
Formel: (C₆H₇Na₃O₁₄S₃)n
MDL: MFCD00081551
CAS-Nummer: 9011-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR