Polyethylenglycol 300, GPR RECTAPUR®

Lieferant: VWR Chemicals

26605.290EA 78 EUR
26605.290 26605.360
Polyethylenglycol 300, GPR RECTAPUR®
Polyethylenglycol 300
Formel: H(OCH₂CH₂)nOH
MDL: MFCD00081839
CAS-Nummer: 25322-68-3

Bestellen

Spezifikation Testergebnisse

Hydroxyl value 356 - 394
pH (5%) 5,0 - 7,0
Viscosity (20°C) 80,0-105,0 mPa.s
Heavy metals (as Pb) max 5 ppm
Ignition residue (SO4) max 0.1 %
Water max 0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR