Polyethylenglycol 4.000, GPR RECTAPUR®

Lieferant: VWR Chemicals

26609.290EA 54.5 EUR
26609.290 26609.460
Polyethylenglycol 4.000, GPR RECTAPUR®
Polyethylenglycol 4.000
Formel: H(OCH₂CH₂)nOH
Lagertemperatur: Raumtemperatur
MDL: MFCD01779612
CAS-Nummer: 25322-68-3

Bestellen

Spezifikation Testergebnisse

Average molecular weight 3500 - 4500
Hydroxyl value 25 - 32
pH (5%) 4,0 - 8,0
Viscosity (20°C, 50% water) 110 - 142 mPa.s
Melting point (clear) about 55°C
Identification (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR