Polyethylenglycol 6.000, GPR RECTAPUR®
Lieferant: VWR Chemicals
26610.290EA
55
EUR
26610.290
26610.360
26610.460
Polyethylenglycol 6.000, GPR RECTAPUR®
Polyethylenglycol 6.000
Formel:
H(OCH₂CH₂)nOH Lagertemperatur: Raumtemperatur |
MDL:
MFCD01779614 CAS-Nummer: 25322-68-3 |
Spezifikation Testergebnisse
Average molecular weight | 5400 - 7000 |
Hydroxyl value | 17 - 21 |
pH (5%) | 4,5 - 7,5 |
Heavy metals (as Pb) | max. 10 ppm |
Ignition residue (SO4) | max. 0.1% |
Water | max. 1.0% |
Solidification point | 55 - 61°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...