Kaliumhydroxid 34% in wässriger Lösung, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: Kalihydrat, Ätzkali

26630.296EA 171 EUR
26630.296
Kaliumhydroxid 34% in wässriger Lösung, AnalaR NORMAPUR® analytisches Reagens
Kaliumhydroxid
Formel: KOH
Molecular Weight: 56,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003553
CAS-Nummer: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay 34.0 - 36.0 %
Ba (Barium) Passes test
Density (20/4) 1.330 - 1.352
Heavy metals (as Pb) Max. 5 ppm
Potassium carbonate Max. 1.0 %
Substances precipitated by NH4OH Max. 100 ppm
Cl (Chloride) Max. 50 ppm
NH4 (Ammonium) Max. 10 ppm
NO3 (Nitrate) Max. 10 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 (Sulphate) Max. 20 ppm
Ca + Mg (as Ca) Max. 100 ppm
Fe (Iron) Max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR