Ammoniumsulfat, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

2674.9025EA 1007.16 EUR
2674.9025 2674.5000 2674-44
Ammoniumsulfat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Ammoniumsulfat
Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
Schmelzpunkt: 336…339 °C
Dichte: 1.78 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Bestellen

Spezifikation Testergebnisse

Assay min. 99.0 %
Arsenic (As) max. 0.3 ppm
Chloride (Cl) max. 3 ppm
Iron (Fe) max. 5 ppm
Heavy Metals (as Pb) max. 5 ppm
Insoluble Matter max. 0.01 %
Nitrate (NO3) max. 5 ppm
pH of 5% Solution at 25°C 5.0 - 6.0
Phosphate (PO4) max. 5 ppm
Residue after Ignition max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR