Ammoniumsulfat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Lieferant: Avantor
Synonyme:
Diammoniumsulfat, di-Ammoniumsulfat
2674.9025EA
1007.16
EUR
2674.9025
2674.5000
2674-44
Ammoniumsulfat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Ammoniumsulfat
Formel:
(NH₄)₂SO₄ Molecular Weight: 132.14 g/mol Schmelzpunkt: 336…339 °C Dichte: 1.78 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003391 CAS-Nummer: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 13,00559 |
Spezifikation Testergebnisse
Assay | min. 99.0 % |
Arsenic (As) | max. 0.3 ppm |
Chloride (Cl) | max. 3 ppm |
Iron (Fe) | max. 5 ppm |
Heavy Metals (as Pb) | max. 5 ppm |
Insoluble Matter | max. 0.01 % |
Nitrate (NO3) | max. 5 ppm |
pH of 5% Solution at 25°C | 5.0 - 6.0 |
Phosphate (PO4) | max. 5 ppm |
Residue after Ignition | max. 0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...