Kaliumchlorat ≥99,0%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

26746.296EA 249 EUR
26746.296
Kaliumchlorat ≥99,0%, AnalaR NORMAPUR® analytisches Reagens
Potassium chlorate
Formel: KClO₃
Molecular Weight: 122.55 g/mol
Siedepunkt: 400
Schmelzpunkt: 356
Dichte: 2.34 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011361
CAS-Nummer: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1 II
Merck Index: 13,07703

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.0 %
Acidity or alkalinity Max. 0.002 meq/g
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Total N (Nitrogen) Max. 100 ppm
BrO3 (Bromate) Max. 150 ppm
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 2 ppm
Ba (Barium) Max. 20 ppm
Ca (Calcium) Max. 20 ppm
Fe (Iron) Max. 5 ppm
Na (Sodium) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR