Kaliumchlorid 99,5-101,0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals

26764.232EA 30.1 EUR
26764.232 26764.298 26764.367 26764.260 26764.460
Kaliumchlorid 99,5-101,0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Potassium chloride
Formel: KCl
Siedepunkt: 1420 °C (1013 hPa)
Schmelzpunkt: 773 °C
Dichte: 1,98 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011360
CAS-Nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Bestellen

Spezifikation Testergebnisse

Assay (calculated on dried substance) 99.5 - 101.0 %
Appearance of Solution S Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Acidity or alkalinity Max. 0.0005 meq/g
pH (5 %) 5.0 - 8.0
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 100 ppm
Loss on drying (105°C) Max. 1.0 %
Mg and alkaline earth metals (as Ca) Max. 0.02 %
Total N (Nitrogen) Max. 40 ppm
Br (Bromide) Max. 0.1 %
I (Iodide) Max. 20 ppm
PO4 (Phosphate) Max. 5 ppm
SO4 (Sulphate) Max. 50 ppm
Al (Aluminium) Max. 10 ppm
As (Arsenic) Max. 0.5 ppm
Ba (Barium) Max. 10 ppm
Ca (Calcium) Max. 10 ppm
Cu (Copper) Max. 2 ppm
Fe (Iron) Max. 2 ppm
Na (Sodium) Max. 0.03 %
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR