tri-Kaliumorthophosphat Trihydrat ≥98,0%, AnalaR NORMAPUR®

Lieferant: VWR Chemicals
Gefahr

26937.290EA 188 EUR
26937.290 26937.460
tri-Kaliumorthophosphat Trihydrat ≥98,0%, AnalaR NORMAPUR®
tri-Kaliumorthophosphat Trihydrat
Formel: K₃O₄P·3H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00151709
CAS-Nummer: 22763-03-7

Bestellen

Spezifikation Testergebnisse

Assay ≥ 98.0 %
Appearance White hygroscopic powder
Identification Passes test
di-Potassium hydrogen phosphate ≤ 2.0 %
Free alkali (as KOH) ≤ 1.0 %
Heavy metals (as Pb) ≤ 20 ppm
Total N (Nitrogen) ≤ 10 ppm
Cl (Chloride) ≤ 30 ppm
As (Arsenic) ≤ 1 ppm
Fe (Iron) ≤ 10 ppm
Na (Sodium) ≤ 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR