Kaliumhydrogenphthalat ≥99,5%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals

26948.146EA 63 EUR
26948.146 26948.237 26948.294 26948.260
Kaliumhydrogenphthalat ≥99,5%, AnalaR NORMAPUR® analytisches Reagens
Kaliumhydrogenphthalat
Formel: HOOCC₆H₄COOK
Schmelzpunkt: 295…300 °C
Dichte: 1,6362 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013070
CAS-Nummer: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calculated on dried substance) Min. 99.5 %
IR Spectrum Passes test
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Loss on drying (105°C) Max. 0.2 %
Sulphur compounds (as SO4) Max. 50 ppm
Cl (Chloride) Max. 20 ppm
Cu (Copper) Max. 2 ppm
Fe (Iron) Max. 5 ppm
Na (Sodium) Max. 100 ppm
Pb (Lead) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR