N,N′-Bis(salicyliden)-1,2-phenylendiamin ≥97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: N,N'-Bis(2-hydroxybenzylidene)-1,2-phenylenediamine, N,N'-Disalicylal-1,2-phenylenediamine, 2,2'-((1,2-Phenylenebis(azanylylidene))bis(methanylylidene))diphenol

269492-5G
269492-5GEA 162 EUR
269492-5G
N,N′-Bis(salicyliden)-1,2-phenylendiamin ≥97%, Sigma-Aldrich®
N,N′-Bis(salicyliden)-1,2-phenylendiamin

N,N'-Bis(salicylidene)-1,2-phenylenediamine,97%

Formel: C₂₀H₁₆N₂O₂
Molecular Weight: 316,36 g/mol
Schmelzpunkt: 163…165 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009995
CAS-Nummer: 3946-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8450f253ec9828601be4e1df099c4d2ff5095791&vid=924603f87e001e824b76e3d9576a9fde622f8b3c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR