Kaliumsorbat, Granulat NF, Ph. Eur., BP

Lieferant: VWR Chemicals
Achtung

26976.291EA 211 EUR
26976.291 26976.462
Kaliumsorbat, Granulat NF, Ph. Eur., BP
Kaliumsorbat

Appearance: White or almost white powder or granules.


Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)

Formel: CH₃CH=CHCH=CHCOOK
Schmelzpunkt: 270 °C
Dichte: 1,36 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00016546
CAS-Nummer: 24634-61-5
EINECS: 246-376-1
Merck Index: 13,07756
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) ≤ 10 ppm
Statement of original manufacturer (*)
Acetone (*) ≤ 200 ppm
Other residual solvents (*) Unlikely by manuf.process
As (Arsenic) Max. 2 ppm
Cd (Cadmium) Max. 1 ppm
Co (Cobalt) Max. 1 ppm
Hg (Mercury) Max. 1 ppm
Ni (Nickel) Max. 1 ppm
Pb (Lead) Max. 2 ppm
V (Vanadium) Max. 1 ppm
Conforms to Ph.Eur./NF/BP Passes test
Ph.Eur./BP specifications Max. 1 ppm
Assay (dried substance) [Ph.Eur./BP] 99.0 - 101.0 %
Identification B Passes test NF
Identification D Passes test Ph.Eur./BP
Appearance of solution Passes test Ph.Eur./BP
Acidity or alkalinity Passes test NF
Aldehydes (calc. as C2H4O) [Ph.Eur./BP] ≤ 0.15 %
Loss on drying (105°C; 3 h) [Ph.Eur./BP] ≤ 1.0 %
NF specifications
Assay (calculated on dried basis) [NF] 98.0 - 102.0 %
Content of potassium [NF] 24.5 - 27.6 %
Identification A Passes test NF
Limit of aldehyde (as acetaldehyde) [NF] ≤ 0.15 %
Loss on drying (105°C; 3 h) [NF] ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR