High purity dNTPs
Lieferant: Cytiva
27-1850-04
27-1860-04
27-1870-04
27-1880-04
27-1850-04EA
478
EUR
27-1850-04
27-1860-04
27-1870-04
27-1880-04
High purity dNTPs
Nukleinsäure-Reagenzien
Nukleotide
dNTPs designed for applications requiring high purity such as amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.
- High purity dNTPs are functionally tested in long PCR to be PCR-qualified, and meet or exceed criteria for high-quality sequencing with thermo sequenase DNA polymerase
- High purity (>98% triphosphate) confirmed by chromatographic analysis and ³¹P nuclear magnetic resonance (NMR) spectroscopy
Bestellinformation: Supplied as solids. For high purity solutions dNTPs, see illustra™ dNTP solutions.