DNA (Deoxyribonucleic acid) (aus Kalbsthymus), beschallt
Lieferant: Cytiva
27-4563-01
27-4563-01EA
613
EUR
27-4563-01
DNA (Deoxyribonucleic acid) (aus Kalbsthymus), beschallt
DNS (Desoxyribonukleinsäure)
Genomic DNA sheared by sonication for use in hybridization experiments.
- Fragmented DNA with a size range from 200-6000 nucleotides
- Purified by phenol extraction and freeze dried
- Tested for absence of nuclease
- Suitable for blocking non-specific binding sites in hybridization experiments to reduce background
DNA isolated from calf thymus is sonicated to fragment the DNA to an average size of 3000 bp.
Lagertemperatur:
Gefrierschrank |
MDL:
MFCD00130922 CAS-Nummer: 9007-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...