Polyribonucleotide poly(I)·poly(C)
Lieferant: Cytiva
27-4732-01
27-4732-01EA
2562
EUR
27-4732-01
Polyribonucleotide poly(I)·poly(C)
Zellkultur-Ergänzungen und -Zusätze
The double-stranded polyribonucleotide poly(I)·poly(C) induces interferon in rabbit cells, and there has been interest in its effectiveness for inducing human interferon in vitro.