illustra™ MicroSpin™ S-300 HR columns
Lieferant: Cytiva
27-5130-01
27-5130-01EA 304 EUR
27-5130-01
illustra™ MicroSpin™ S-300 HR columns
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
illustra™ MicroSpin™ S-300 HR columns designed for rapid purification of PCR products (>100 bp) from unincorporated primers (<20-mers) and nucleotides using spin-column chromatography.
  • Accommodates 25 to 50 μl for post-PCR clean-up prior to sequencing
  • Useful for purification of alkaline-denatured plasmid DNA prior to sequencing
  • Prepacked for convenience with Sephacryl™ S-300 HR pre-equilibrated in TE buffer
  • Ready to use, requiring less than 4 minutes from sample application to collection of purified product
  • Tested in nickase, single- and double-stranded exonuclease and RNase assays