Mineralöl, weiß stabilisiert USP, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Stabilisierung:
Stabilisiert mit Vitamin E 10-100 ppm
Veränderungsmanagement-Kategorie= R
2705-01EA
59.86
EUR
2705-01
Mineralöl, weiß stabilisiert USP, J.T.Baker®
Mineralöl
Siedepunkt:
260 °C Dichte: 0,85 g/cm³ (20 °C) Flammpunkt: 135 °C (geschlossener Tiegel) |
MDL:
MFCD00131611 CAS-Nummer: 8012-95-1 EINECS: 232-384-2 Merck Index: 13,07093 |
Spezifikation Testergebnisse
GMP Manufactured Product | |
Meets U.S.P Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | Passes Test |
USP - Acidity | Passes Test |
USP - Limit of Polycyclic Aromatic Hydrocarbons | Passes Test |
USP - Limit of Sulfur Compounds | Passes Test |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Specific Gravity at 25°/25°C | 0.845 - 0.905 |
USP - Viscosity at 40 C, mm₂/s | 34.5 - 150.0 |
USP - Readily Carbonizable Substances | Passes Test |
USP - Solid Paraffin | Passes Test |
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...