L(+)-Kaliumnatriumtartrat Tetrahydrat, TECHNICAL

Lieferant: VWR Chemicals

27062.290EA 60.5 EUR
27062.290
L(+)-Kaliumnatriumtartrat Tetrahydrat, TECHNICAL
L(+)-Kaliumnatriumtartrat Tetrahydrat
Formel: KOOC(CHOH)₂COONa·4H₂O
Schmelzpunkt: 70…80 °C
Dichte: ∼1,767 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150989
CAS-Nummer: 6381-59-5
EINECS: 206-156-8
REACH: 01-2119524666-34
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR