Wasser, CHROMASOLV™ HPLC, Riedel-de Haën™

Lieferant: Honeywell Chemicals

Synonyme: Wasserstoffoxid, Hydrogenoxid

270733-1L 270733-2.5L 270733-4L 270733-6X1L
270733-1LEA 69.78 EUR
270733-1L 270733-2.5L 270733-4L 270733-6X1L
Wasser, CHROMASOLV™ HPLC, Riedel-de Haën™
Wasser

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100 °C
Schmelzpunkt: 0 °C
Dichte: 0.9971 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Non-volatile matter Max. 0.0003 %
Absorbance at 400 nm Max. 0.005
Absorbance at 250 nm Max. 0.005
Absorbance at 210 nm Max. 0.01
Absorbance at 205 nm Max. 0.01
HPLC Gradient at 254 nm Max. 2 mAU
HPLC Gradient at 210 nm Max. 10 mAU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR