N-Octadecanoyl nile blue, Selectophore™ Ionophor, Supelco®

Lieferant: Merck

Synonyme: Chromoionophore I

M-Clarity™ Qualitätsstufe = MQ100

27086-100MG-F 27086-10MG-F
27086-100MG-FEA 453 EUR
27086-100MG-F 27086-10MG-F
N-Octadecanoyl nile blue, Selectophore™ Ionophor, Supelco®
N-Octadecanoyl nile blue

Chromoionophore I (ETH 5294, 3-octadecanoylimino-7-diethylamino-l,2-benzophenoxazine) is a H+-selective neutral chromoionophore, which is lipophilic in nature and is a highly basic dye molecule.


Chromoionophore I may be used for the test zone preparation in the multianalyte sensor, during environmental monitoring, using photographic digital camera.


Assay of K+ and of Ca2+ activity with solvent polymeric optode membrane based on ETH 5294 and potassium ionophore I (valinomycin).


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₃₈H₅₃N₃O₂
CAS-Nummer: 125829-24-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71b5fd7f2cdb48d3f1697268808fb6795fdbb382&vid=ab4089d035b92e4dcf9e285400024ad4f4e1a005&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR