N-Octadecanoyl nile blue, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
Synonyme:
Chromoionophore I
M-Clarity™ Qualitätsstufe = MQ100
Chromoionophore I (ETH 5294, 3-octadecanoylimino-7-diethylamino-l,2-benzophenoxazine) is a H+-selective neutral chromoionophore, which is lipophilic in nature and is a highly basic dye molecule.
Chromoionophore I may be used for the test zone preparation in the multianalyte sensor, during environmental monitoring, using photographic digital camera.
Assay of K+ and of Ca2+ activity with solvent polymeric optode membrane based on ETH 5294 and potassium ionophore I (valinomycin).
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₃₈H₅₃N₃O₂ |
CAS-Nummer:
125829-24-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...