Adapters, pig receiver, Quickfit®
Lieferant: SciLabware
RA5/11/SC
271-0013EA 143 EUR
271-0013
Adapters, pig receiver, Quickfit®
Destillationsbauteile Destilliervorstöße
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Allows up to three flasks to be connected to a condenser outlet. Useful for collecting distillate fractions. By slightly rotating the adapter a new flask is presented to the distillate. With side-arm for connection to a vacuum supply.