Säulenkopf mit Ventilhahn
Lieferant: Lenz Laborglas GmbH & CO.KG
05170144
271-3215EA 998 EUR
271-3215
Säulenkopf mit Ventilhahn
Destillationsbauteile Destillieraufsätze
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With PTFE valve for easy and accurate reflux adjustment, with thermometer connection NS 14/23 for thermometers with 55 mm built-in length.