2,5-Dibromo-6-isopropyl-3-methyl-1,4-benzoquinone, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 2,5-Dibromo-3-methyl-6-isopropyl-p-benzoquinone, Dibromothymoquinone

271993-250MG
271993-250MGEA 156 EUR
271993-250MG
2,5-Dibromo-6-isopropyl-3-methyl-1,4-benzoquinone, Sigma-Aldrich®
2,5-Dibromo-6-isopropyl-3-methyl-1,4-benzoquinone

2, 5-Dibromo-6-isopropyl-3-methyl-1, 4-benzoquinone has been used as a metabolic inhibitor of violaxanthin (V) cycle of Pelvetia canaliculata and is used as a photosynthetic electron transport chain (pETC) inhibitor.

Formel: CHBr₂O₂
Molecular Weight: 321,99 g/mol
Schmelzpunkt: 70…72 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009967
CAS-Nummer: 29096-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=974cad1e82c54f302879418eb6a70e483e4ca401&vid=60d0c01824f153d5335dc99a7bbe86ae50c16d7d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR