Glass cutter
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
K-1915
272-9015EA 32.9 EUR
272-9015
Glass cutter
Schneidewerkzeuge
Wooden handle with special steel wheels.
Glass cutter with six special steel wheels for cutting glass plates, TL-plates, microslides, etc.