Astrazon Orange G, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Basic Orange 21

274216-100G
274216-100GEA 215 EUR
274216-100G
Astrazon Orange G, Sigma-Aldrich®
Astrazon Orange G

Metachromatic dye

Formel: C₂₂H₂₃ClN₂
Molecular Weight: 350.88 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011947
CAS-Nummer: 3056-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f34fac651b1e86924b8029c30aedcefadfa466ff&vid=525848d4f43a4bd2f15955adcb83c8675339fcc3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR