Lacmoid, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Resorcinol Blue

M-Clarity™ Qualitätsstufe = MQ100

274720-10G 274720-25G
274720-10GEA 122 EUR
274720-10G 274720-25G
Lacmoid, Sigma-Aldrich®
Lacmoid
Lagertemperatur: Room Temperature
MDL: MFCD00152579
CAS-Nummer: 42249-61-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcc9a87d6bdd641af48248b1a15d98b55bb4cf89&vid=46529c95f3b2fda9b025a2e68ec6fc835131e20e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR