D(+)-Saccharose, AnalaR NORMAPUR® zA

Lieferant: VWR Chemicals

Synonyme: Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker

27480.294EA 112 EUR
27480.294 27480.260 27480.360
D(+)-Saccharose, AnalaR NORMAPUR® zA
D(+)-Saccharose
Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342.3 g/mol
Schmelzpunkt: 169…170 °C
Dichte: 1.29 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006626
CAS-Nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Acidity Max. 0.005 meq/g
Specific optical rotation (26 %; water) 66.4 - 66.8 °
Glucose Max. 0.05 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 80 ppm
Insolubility in water Max. 30 ppm
Loss on drying (100°C) Max. 0.05 %
Reducing sugars (calculated as C6H12O6) Max. 0.1 %
Cl (Chloride) Max. 5 ppm
N (Nitrogen) Max. 20 ppm
SO3 (as SO2) Max. 15 ppm
SO4 (Sulphate) Max. 20 ppm
Ba (Barium) Max. 0.02 %
Ca (Calcium) Max. 10 ppm
Cd (Cadmium) Max. 0.05 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.5 ppm
Ni (Nickel) Max. 0.1 ppm
Pb (Lead) Max. 0.5 ppm
Zn (Zinc) Max. 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR