(S)-(-)-β-Citronellol, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

27483-1ML-F 27483-5ML-F
27483-1ML-FEA 80.2 EUR
27483-1ML-F 27483-5ML-F
(S)-(-)-β-Citronellol, Supelco®
(S)-(-)-β-Citronellol
Formel: C₁₀H₂₀O
Dichte: 0,855 g/cm³ (20 °C)
MDL: MFCD00063214
CAS-Nummer: 7540-51-4
EINECS: 231-415-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR