D(+)-Saccharose Ph. Eur.

Lieferant: VWR Chemicals

Synonyme: Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker

27483.294EA 63.3 EUR
27483.294 27483.363 27483.465
D(+)-Saccharose Ph. Eur.
D(+)-Saccharose

Bitte beachten Not for parenteral use.

Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342.3 g/mol
Schmelzpunkt: 169…170 °C
Dichte: 1.29 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006626
CAS-Nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Conforms (see CoA/CoS)
Identification (IR) Passes test
Appearance of solution Passes test
Conductivity (20°C; 31,3 %) Max. 35 µS/cm
Specific optical rotation (26 %; water) 66.3 - 67 °
Colour value Max. 45
Reducing sugars Passes test
SO3 (as SO2) Max. 10 ppm
Loss on drying (105°C; 3 h) Max. 0.1 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR