Salicylaldoxim
Lieferant: VWR Chemicals
Synonyme:
2-Hydroxybenzaldehyde oxime
27491.188EA
0
EUR
27491.188
Salicylaldoxim
Salicylaldoxim
Formel:
C₇H₇NO₂ Molecular Weight: 137,14 g/mol |
MDL:
MFCD00002120 CAS-Nummer: 94-67-7 EINECS: 202-353-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...