3-Methoxydiphenylamin 98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 3-Methoxy-N-phenylaniline

M-Clarity™ Qualitätsstufe = MQ100

275980-25G
275980-25GEA 282 EUR
275980-25G
3-Methoxydiphenylamin 98%, Sigma-Aldrich®
3-Methoxydiphenylamin

3-Methoxydiphenylamine can be used in the preparation of 3- methoxydiphenylamine-4-diazonium salts (MDP-4-DS) and the corresponding diazoresins.[1] Diazonium salts and diazoresins find applications in dyes, stains, indicators, general laboratory use, and industrial applications.

Formel: C₁₃H₁₃NO
Molecular Weight: 199,25 g/mol
Schmelzpunkt: 72,..74 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00008383
CAS-Nummer: 101-16-6
EINECS: 202-921-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1192e235877d7a27acbb22efba439fcc9dbaf893&vid=1192e235877d7a27acbb22efba439fcc9dbaf893&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to brown powder, crystals, crystalline Powder and/or chunks
Infrared spectrum Conforms to structure
Elemental analysis Carbon 76.4 - 80.3%, Nitrogen 6.7 - 7.3%
Gas liquid 97.5% (minimum)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR