3-Noradamantanecarboxylic acid 97%

Lieferant: SIGMA ALDRICH MICROSCOPY

276235-1G
276235-1GEA 318 EUR
276235-1G
3-Noradamantanecarboxylic acid 97%
3-Noradamantanecarboxylic acid

Building block for 'Anti-Bredt' strained molecules.

Formel: C₁₀H₁₄O₂
Molecular Weight: 166.22 g/mol
Schmelzpunkt: 105...107 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00213463
CAS-Nummer: 16200-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12a1a44f4d19fe925ea132f5ab73b750d35f6e34&vid=a58a8b0fe36d4812de1edb67926c459b788d51ec&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Purity (Titration by NAOH) 96.5 - 103.5%
GC (area %) ≥96.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR