Methylviolett, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Fleisher's Indicator Solution
277339-1L
277339-1LEA
116
EUR
277339-1L
Methylviolett, Sigma-Aldrich®
Methylviolett
Methyl Purple,in H2O (Methyl Purple indicator solution)
Formel:
C₆₇H₆₂N₈Na₄O₁₄S₃ Molecular Weight: 1391,41 g/mol Schmelzpunkt: 0 °C Dichte: 1,001 g/mL (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070843 CAS-Nummer: 1340-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...