Natriumbromid ≥99%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals

Synonyme: Bromnatrium

27739.298EA 190 EUR
27739.298
Natriumbromid ≥99%, AnalaR NORMAPUR® analytisches Reagens
Natriumbromid
Formel: NaBr
Molecular Weight: 102,89 g/mol
Siedepunkt: 1396 °C (1013 hPa)
Schmelzpunkt: 753 °C
Dichte: 3,2 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003475
CAS-Nummer: 7647-15-6
EINECS: 231-599-9
REACH: 01-2119490106-41
Merck Index: 13,08666

Bestellen

Spezifikation Testergebnisse

Assay (calculated on dried substance) Min. 99 %
Acidity Max. 0.002 meq/g
Alkalinity Max. 0.004 meq/g
Heavy metals (as Pb) Max. 10 ppm
Insolubility in water Max. 50 ppm
Loss on drying (150°C) Max. 2 %
BrO3 (Bromate) Max. 5 ppm
Cl (Chloride) Max. 0.3 %
NH4 (Ammonium) Max. 20 ppm
SO4 (Sulphate) Max. 100 ppm
As (Arsenic) Max. 2 ppm
Ca (Calcium) Max. 0.02 %
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 0.1 %
Mg (Magnesium) Max. 20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR