Natriumhydrogencarbonat 99,7-100,3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals

27778.236EA 28.4 EUR
27778.236 27778.293 27778.360 27778.260 27778.460
Natriumhydrogencarbonat 99,7-100,3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Natriumhydrogencarbonat
Formel: NaHCO₃
Schmelzpunkt: 270 °C
Dichte: 2.159 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003528
CAS-Nummer: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calc. on dried substance) 99.7 - 101,0 %
Appearance of solution Passes test Ph.Eur.
Carbonates Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Identification C Passes test Ph.Eur.
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 0.015 %
Loss on drying Max. 0.2 %
Reducing substances (as I) Max. 65 ppm
Total N (Nitrogen) Max. 5 ppm
Total S (as SO4) Max. 30 ppm
Cl (Chloride) Max. 50 ppm
NH4 (Ammonium) Max. 5 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 (Sulphate) Max. 20 ppm
As (Arsenic) Max. 2 ppm
Ca (Calcium) Max. 100 ppm
Cu (Copper) Max. 2 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 50 ppm
Pb (Lead) Max. 5 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR