Methyl-3-amino-5,6-dichlorpyrazin-2-carboxylat 97%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
M-Clarity™ Qualitätsstufe = MQ200
277878-5G
277878-5GEA
453
EUR
277878-5G
Methyl-3-amino-5,6-dichlorpyrazin-2-carboxylat 97%, Sigma-Aldrich®
Methyl-3-amino-5,6-dichlorpyrazin-2-carboxylat
Formel:
C₆H₅Cl₂N₃O₂ Molecular Weight: 222.03 g/mol Schmelzpunkt: 230...233 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00010431 CAS-Nummer: 1458-18-0 EINECS: 215-947-7 |
|
Spezifikation Testergebnisse
Appearance (Color) | Faint Brown to Dark Brown |
Appearance (Form) | Powder or Crystal or Chunk(s) |
Infrared spectrum | Conforms to Structure |
Titration by AgNO3 | 96.5 - 103.5 % |
Purity (HPLC) | ≥96.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...