Carbopol® 940

Lieferant: Thermo Fisher Scientific

278380000
278380000.EA 0 EUR
278380000.
Carbopol® 940
Carbomer 940

Appearance: White Powder

Lagertemperatur: Raumtemperatur
MDL: MFCD01941675
CAS-Nummer: 9003-01-4

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
Heavy metals ≤10 ppm (Hg, Pb, As, Sb)
Viscosity 40000 to 60000 cP (at 25°C) (Brookfield, 20 rpm) (neutralized soln., 0.5% dispersion)
Clarity of solution ≥85 % transmission (neutralized soln., 420 nm, 0.5% dispersion)
Residual solvents ≤0.5 % Benzene


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR