Natriumformiat ≥98%, GPR RECTAPUR®

Lieferant: VWR Chemicals

Synonyme: Ameisensäure Natriumsalz

27869.297EA 93.5 EUR
27869.297
Natriumformiat ≥98%, GPR RECTAPUR®
Natriumformiat

METHOXYPENTANE™ is a novel high quality ether solvent developed by INEOS Oligomers for demanding applications in the manufacture of high value fine chemicals, advanced life science intermediates, API’s, and high-tech chemicals. Methoxypentane™ is ultra-pure with a 2-methoxy-2-methyl butane content typically exceeding 99.5 wt.-%.


METHOXYPENTANE™ offers a range of unique solvent properties which helps to support user creativity in the identification of new applications for the product. The novel solvent enables researchers in academic and industrial settings to invent and design new synthesis processes and purification procedures. Furthermore, it allows process development professionals in industry to establish efficient manufacturing routes and production managers to achieve and improve selectivity and product yield while reducing waste.

Formel: HCO₂Na
Molecular Weight: 68,01 g/mol
Schmelzpunkt: 257,31 °C
Dichte: 1,919 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013101
CAS-Nummer: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 13,08694

Bestellen

Spezifikation Testergebnisse

Assay Min. 98 %
Heavy metals (as Pb) Max. 5 ppm
Cl (Chloride) Max. 20 ppm
NH4 (Ammonium) Max. 20 ppm
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR