Natriumnitrat ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens

Lieferant: VWR Chemicals
Achtung

27955.238EA 29.6 EUR
27955.238 27955.295 27955.460
Natriumnitrat ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens
Natriumnitrat
Formel: NaNO₃
Siedepunkt: 380 °C (1013 hPa)
Schmelzpunkt: 308 °C
Dichte: 2,26 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011119
CAS-Nummer: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 13,08720

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.5 %
pH (25°C; 5 %) 5.5 - 8.3
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 5 ppm
IO3 (Iodate) Max. 5 ppm
NH4 (Ammonium) Max. 20 ppm
NO2 (Nitrite) Max. 10 ppm
PO4 (Phosphate) Max. 5 ppm
SO4 (Sulphate) Max. 30 ppm
Ca (Calcium) Max. 20 ppm
Fe (Iron) Max. 3 ppm
Mg (Magnesium) Max. 20 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR