Nitroprussid-Natrium Dihydrat 99,0-102,0%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: Dinatriumpentacyanonitrosylferrat(III) Dihydrat, Natriumnitrosopentacyanoferrat(III) Dihydrat, di-Natriumnitrosylpentacyanoferrat(III) Dihydrat, Natriumnitroprussiat Dihydrat, di-Natriumpentacyanonitrosylferrat(III) Dihydrat

27966.180EA 126 EUR
27966.180
Nitroprussid-Natrium Dihydrat 99,0-102,0%, AnalaR NORMAPUR® analytisches Reagens
Nitroprussid-Natrium Dihydrat
Formel: Na₂[Fe(CN)₅NO]·2H₂O
Molecular Weight: 297,95 g/mol
Dichte: 1,72 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149192
CAS-Nummer: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,III
Merck Index: 13,08722

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 102.0 %
Hexacyanoferrate (II) (as Fe(CN)6) Max. 0.02 %
Hexacyanoferrate (III) (as Fe(CN)6) Max. 0.02 %
Insolubility in methanol Max. 0.5 %
Insoluble matter Max. 100 ppm
Cl (Chloride) Max. 100 ppm
SO4 (Sulphate) Max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR