Ab buffer kit
Lieferant: Cytiva
28-9030-59
28-9030-59EA
174
EUR
28-9030-59
Ab buffer kit
Puffer
Puffer für die Antikörperreinigung
Ab buffer kit contains prepared buffer concentrates for binding, washing, and elution of IgG according to recommended protocols.
The kit also includes neutralising buffer. Ab buffer kit eliminates time consuming buffer preparation, which allows fast, reproducible, and convenient purification work.
Lieferumfang: Includes: 1×50 ml binding buffer (10X stock solution), 1×15 ml elution buffer (10X stock solution), and 1×25 ml neutralising buffer.