Cation exchange chromatography column, HiPrep™ IEX SP Sepharose™ XL
Lieferant: Cytiva
28-9365-40
28-9365-40EA 745 EUR
28-9365-40
Cation exchange chromatography column, HiPrep™ IEX SP Sepharose™ XL
Chromatographiesäulen
HiPrep™ SP XL 16/10 is a prepacked, ready to use column for ion exchange chromatography.
  • Provide high loading capacity and fast purifications
  • Optimized for reliable, reproducible separations
  • Compatible with simple pump-based configurations and chromatography systems such as ÄKTA design
  • Excellent choice for rapid enrichment during initial capture of proteins from start material
  • Strong Sulfopropyl (SP) cation exchanger
It is prepacked with Q Sepharose™ XL for inital capture of proteins and other biomoleculs with ion exchange (IEX) chromatography.