3,3'-Diaminobenzidin Tetrahydrochlorid (DAB)

Lieferant: Merck Millipore (Oncogene)

Synonyme: 3,3',4,4'-Biphenyltetramine tetrahydrochloride, DAB HCl

281751-10
281751-10EA 182 EUR
281751-10
3,3'-Diaminobenzidin Tetrahydrochlorid (DAB)
3,3'-Diaminobenzidin Tetrahydrochlorid
Formel: C₁₂H₁₈Cl₄N₄
Molecular Weight: 360,11 g/mol
MDL: MFCD00012969
CAS-Nummer: 7411-49-6
EINECS: 231-018-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR