Rhodamin B Isothiocyanat Gemisch aus Isomeren, Pulver, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: RBITC

M-Clarity™ Qualitätsstufe = MQ200

283924-100MG 283924-500MG
283924-100MGEA 116 EUR
283924-100MG 283924-500MG
Rhodamin B Isothiocyanat Gemisch aus Isomeren, Pulver, Sigma-Aldrich®
Rhodamin B Isothiocyanat

Rhodamine B isothiocyanate (RBITC) is a fluorescent dye. It acts as an anterograde and retrograde marker of retinal neurons in the adult rat

Formel: C₂₉H₃₀ClN₃O₃S
Molecular Weight: 536.08 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00136007
CAS-Nummer: 36877-69-7
EINECS: 253-248-9
UN: 3335
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aca91c5f700e3bc22ffcf0ca3da21d54ee259f25&vid=4a3b31c5db8544d6ffabb4b7e6da9152974d4d41&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR