D(+)-Glucose, wasserfrei 97,5-102,0% USP, Ph. Eur.

Lieferant: VWR Chemicals

284504SEA 42 EUR
284504S 284508W 28450BF
D(+)-Glucose, wasserfrei 97,5-102,0% USP, Ph. Eur.
D(+)-Glucose

Not for use as Active Pharmaceutical Ingredient (API)


Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)

Formel: C₆H₁₂O₆
Schmelzpunkt: 146 °C
Dichte: 1,54 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance: 97,5 - 102,0 %
White or almost white crystalline powder Passes test
Heavy metals (as Pb) Max. 5 μg/g
Residual solvents Unlikely by manuf.process
Residues of metal catalysts or reagents Conforms to EMEA/CHMP/SWP/4446/2000
As (Arsenic) Max. 2 ppm
Cd (Cadmium) Max. 1 ppm
Co (Cobalt) Max. 1 ppm
Hg (Mercury) Max. 2 ppm
Ni (Nickel) Max. 1 ppm
Pb (Lead) Max. 2 ppm
V (Vanadium) Max. 1 ppm
Conforms to Ph.Eur./USP Passes test
Ph.Eur. specifications: Max. 0,5 %
Assay (on anhydrous) [Ph.Eur.] 97.5 --> 102.0 %
Identification A [Ph.Eur.] 52.5 --> 53.3 °
Identification B Passes test USP
Identification E Passes test Ph.Eur.
Appearance of solution Passes test Ph.Eur.
Conductivity [Ph.Eur.] Max. 20 μS/cm
Maltose and isomaltose [Ph.Eur.] Max. 0.4 %
Maltotriose [Ph.Eur.] Max. 0.2 %
Fructose [Ph.Eur.] Max. 0.15 %
Unspecified impur. (for each) [Ph.Eur.] Max. 0.10 %
Total impurities [Ph.Eur.] Max. 0.5 %
Disregard limit [Ph.Eur.] Max. 0.05 %
Dextrin Passes test USP
Soluble starch, sulfite [Ph.Eur.] Max. 15 ppm
Water [Ph.Eur.] Max. 1.0 %
USP specifications: Conforms to EMEA/CHMP/SWP/4446/2000
Assay (calculated on anhydrous) [USP] 97.5 --> 102.0 %
Identification A Passes test USP
Identification C [USP] Max. 1.0 %
Maltose and isomaltose [USP] Max. 0.4 %
Maltotriose [USP] Max. 0.2 %
Fructose [USP] Max. 0.15 %
Unspecified impurities (for each) [USP] Max. 0.10 %
Total impurities [USP] Max. 0.5 %
Disregard limit [USP] Max. 0.05 %
Colour and clarity of solution Passes test USP
Conductivity (25°C) [USP] Max. 20 μS/cm
Soluble starch, sulfites [USP] Max. 15 ppm
For info:


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR