Buffer concentrate, citrate phosphate, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
2851-100ML
2851-100MLEA
103
EUR
2851-100ML
Buffer concentrate, citrate phosphate, Sigma-Aldrich®
Puffer
Biologische Puffer
These buffers are used to elute adult hemoglobin by the procedure of Oski and Naiman, a modification of the Kleihauer Betke procedure for the demonstration of fetal hemoglobin.