Methylenblau Hydrat ≥95% (Trockenmasse), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 3,7-Bis(dimethylamino)phenothiaziniumchlorid Hydrat, Methylthioniniumchlorid Hydrat, N,N,N′,N′-Tetramethylthioninchlorid Hydrat

M-Clarity™ Qualitätsstufe = MQ200

28514-100G 28514-1KG
28514-100GEA 68.9 EUR
28514-100G 28514-1KG
Methylenblau Hydrat ≥95% (Trockenmasse), Sigma-Aldrich®
Methylenblau Hydrat
Formel: C₁₆H₁₈ClN₃S·xH₂O
Molecular Weight: 319.86 g/mol (anhy)
MDL: MFCD00150006
CAS-Nummer: 122965-43-9
EINECS: 200-515-2

Bestellen

Spezifikation Testergebnisse

Titration ≥95%
Titration (method) Argentometric after oxygen combustion, calculated to the dried substance
Solubility Insoluble in ethanol ≤1%
Loss on drying ≤ 22% (105 c)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR