Acid hematoxylin solution, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung
2852-100ML
2852-100MLEA 104 EUR
2852-100ML
Acid hematoxylin solution, Sigma-Aldrich®
Acid hematoxylin solution
Nuclear counterstain. Included as a component in the Sigma Fetal Hemoglobin Procedure 285.
IVDLogo